siRNA Details
| Gene Symbol: FAM185A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026879.1 | 1, 2 | 465 | |||
| GenBank mRNA | AK054745.1 | - | 1, 2 | 375 | ||
| AK297455.1 | - | 2, 3 | 186 | |||
| DB192367.1 | - | 1, 2 | 465 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM185A| Gene Symbol: FAM185A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026879.1 | 1, 2 | 465 | |||
| GenBank mRNA | AK054745.1 | - | 1, 2 | 375 | ||
| AK297455.1 | - | 2, 3 | 186 | |||
| DB192367.1 | - | 1, 2 | 465 | |||