siRNA Details
| Gene Symbol: LINC00879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015400.1 | 8 | 1005 | |||
| GenBank mRNA | BC038721.1 | - | 8 | 1008 | ||
| BC048119.1 | - | 8 | 1006 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB515319.2 | - | 5 | 658 | Chr. 3 - 94980378 - 94991324 [+] on Build GRCh38 | |

LINC00879
Genomic Map