siRNA Details
| Gene Symbol: LINC00879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015400.1 | 7 | 847 | |||
| GenBank mRNA | BC038721.1 | - | 7 | 850 | ||
| BC048119.1 | - | 7 | 848 | |||
| BC104171.1 | - | 6 | 682 | |||
| BC104172.1 | - | 4 | 244 | |||
| BC115359.1 | - | 7 | 766 | |||
| BC115360.1 | - | 7 | 799 | |||

LINC00879
Genomic Map