siRNA Details
| Gene Symbol: LINC00879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015400.1 | 4 | 456 | |||
| GenBank mRNA | BC038721.1 | - | 4 | 459 | ||
| BC048119.1 | - | 4 | 457 | |||
| BC104171.1 | - | 3 | 291 | |||
| BC115359.1 | - | 4 | 375 | |||
| BC115360.1 | - | 4 | 408 | |||

LINC00879
Genomic Map