siRNA Details
| Gene Symbol: LINC00957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015401.2 | 3 | 1699 | |||
| GenBank mRNA | BC081571.1 | - | 3 | 1577 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00957| Gene Symbol: LINC00957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015401.2 | 3 | 1699 | |||
| GenBank mRNA | BC081571.1 | - | 3 | 1577 | ||