siRNA Details
| Gene Symbol: SNHG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095849.1 | - | 4 | 1869 | ||
| Gene Symbol: SNORD22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_000008.2 | 1 | 19 | |||

SNHG1
Genomic Map