siRNA Details
| Gene Symbol: CASC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026939.1 | 2 | 310 | |||
| NR_026940.1 | 2 | 310 | ||||
| NR_026941.1 | 2 | 310 | ||||
| GenBank mRNA | AJ344228.1 | - | 2 | 123 | ||
| AJ535620.1 | - | 2 | 123 | |||
| AJ535621.1 | - | 2 | 123 | |||
| AJ535622.1 | - | 2 | 137 | |||
| AK055184.1 | - | 2 | 137 | |||
| AY226450.1 | - | 2 | 126 | |||
| AY226451.1 | - | 2 | 126 | |||
| BC036393.1 | - | 2 | 312 | |||
| BC112306.1 | - | 1 | 34 | |||
| BC112308.1 | - | 1 | 34 | |||
| BC112326.1 | - | 2 | 130 | |||

CASC2
Genomic Map