siRNA Details
| Gene Symbol: RAB9BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_000039.2 | 1 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB9BP1| Gene Symbol: RAB9BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_000039.2 | 1 | 365 | |||