siRNA Details
| Gene Symbol: RRN3P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027460.1 | 6 | 1377 | |||
| GenBank mRNA | AK055742.1 | - | 6 | 1377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RRN3P3| Gene Symbol: RRN3P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027460.1 | 6 | 1377 | |||
| GenBank mRNA | AK055742.1 | - | 6 | 1377 | ||