siRNA Details
| Gene Symbol: GSTTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003081.1 | 4 | 695 | |||
| GenBank mRNA | AL050257.1 | - | 4 | 694 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSTTP1| Gene Symbol: GSTTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003081.1 | 4 | 695 | |||
| GenBank mRNA | AL050257.1 | - | 4 | 694 | ||