siRNA Details
| Gene Symbol: PISRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027070.1 | 1 | 87 | |||
| GenBank mRNA | AI204197.1 | - | 1 | 87 | ||
| AW268472.1 | - | 1 | 87 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PISRT1| Gene Symbol: PISRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027070.1 | 1 | 87 | |||
| GenBank mRNA | AI204197.1 | - | 1 | 87 | ||
| AW268472.1 | - | 1 | 87 | |||