siRNA Details
| Gene Symbol: GUSBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024448.2 | 8 | 2480 | |||
| GenBank mRNA | AK056875.1 | - | 8 | 2410 | ||
| AK091837.1 | - | 7 | 2218 | |||
| BX538181.1 | - | 7 | 2205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUSBP11| Gene Symbol: GUSBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024448.2 | 8 | 2480 | |||
| GenBank mRNA | AK056875.1 | - | 8 | 2410 | ||
| AK091837.1 | - | 7 | 2218 | |||
| BX538181.1 | - | 7 | 2205 | |||