siRNA Details
| Gene Symbol: GUSBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024448.2 | 5 | 858 | |||
| GenBank mRNA | AK056875.1 | - | 5 | 791 | ||
| AK091837.1 | - | 5 | 716 | |||
| AK300413.1 | - | 5 | 760 | |||
| BC063045.1 | - | 5 | 621 | |||
| BX538181.1 | - | 5 | 706 | |||

GUSBP11
Genomic Map