siRNA Details
| Gene Symbol: DGCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024159.1 | 1 | 2180 | |||
| GenBank mRNA | BC036821.1 | - | 1 | 2191 | ||
| L77571.1 | - | 1 | 2078 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DGCR9| Gene Symbol: DGCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024159.1 | 1 | 2180 | |||
| GenBank mRNA | BC036821.1 | - | 1 | 2191 | ||
| L77571.1 | - | 1 | 2078 | |||