siRNA Details
| Gene Symbol: LINC01018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024423.2 | 5 | 1197 | |||
| NR_024424.2 | 3 | 1211 | ||||
| GenBank mRNA | AK125283.1 | - | 5 | 1164 | ||
| BC043253.2 | - | 3 | 900 | |||
| BC106918.2 | - | 3 | 1195 | |||
| BC106919.1 | - | 3 | 1198 | |||

LINC01018
Genomic Map