siRNA Details
| Gene Symbol: LOC148696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026817.1 | 1 | 487 | |||
| GenBank mRNA | AK125868.1 | - | 1 | 488 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC148696| Gene Symbol: LOC148696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026817.1 | 1 | 487 | |||
| GenBank mRNA | AK125868.1 | - | 1 | 488 | ||