siRNA Details
| Gene Symbol: UBA6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015439.1 | 2 | 1281 | |||
| GenBank mRNA | AK093682.1 | - | 2 | 1198 | ||
| AK129519.1 | - | 2 | 1226 | |||
| BC043376.1 | - | 3 | 435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBA6-AS1| Gene Symbol: UBA6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015439.1 | 2 | 1281 | |||
| GenBank mRNA | AK093682.1 | - | 2 | 1198 | ||
| AK129519.1 | - | 2 | 1226 | |||
| BC043376.1 | - | 3 | 435 | |||