siRNA Details
| Gene Symbol: UBA6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015439.1 | 2 | 1627 | |||
| GenBank mRNA | AK093682.1 | - | 2 | 1544 | ||
| AK129519.1 | - | 2 | 1572 | |||
| BC043376.1 | - | 3 | 781 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI756252.1 | - | 3 | 780 | Chr. 4 - 67701282 - 67721915 [+] on Build GRCh38 | |

UBA6-AS1
Genomic Map