siRNA Details
| Gene Symbol: LOC148709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002929.2 | 3 | 1639 | |||
| GenBank mRNA | AK055809.1 | - | 3 | 1639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC148709| Gene Symbol: LOC148709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002929.2 | 3 | 1639 | |||
| GenBank mRNA | AK055809.1 | - | 3 | 1639 | ||