siRNA Details
| Gene Symbol: ANKRD26P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026556.1 | 11 | 1941 | |||
| GenBank mRNA | BC070117.1 | - | 11 | 1941 | ||
| CD000265.1 | - | 3 | 475 | |||
| CR749347.1 | - | 3 | 437 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD26P1| Gene Symbol: ANKRD26P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026556.1 | 11 | 1941 | |||
| GenBank mRNA | BC070117.1 | - | 11 | 1941 | ||
| CD000265.1 | - | 3 | 475 | |||
| CR749347.1 | - | 3 | 437 | |||