siRNA Details
Gene Symbol: ANKRD26P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026556.1 | 17 | 3752 | |||
GenBank mRNA | AK125968.1 | - | 5 | 847 | ||
BX648636.1 | - | 7 | 1535 | |||
CR749347.1 | - | 10 | 2375 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ANKRD26P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026556.1 | 17 | 3752 | |||
GenBank mRNA | AK125968.1 | - | 5 | 847 | ||
BX648636.1 | - | 7 | 1535 | |||
CR749347.1 | - | 10 | 2375 |