siRNA Details
| Gene Symbol: ANKRD26P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026556.1 | 17 | 5990 | |||
| GenBank mRNA | AK125968.1 | - | 5 | 3086 | ||
| BX648636.1 | - | 7 | 3773 | |||
| CR749347.1 | - | 10 | 4613 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD26P1| Gene Symbol: ANKRD26P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026556.1 | 17 | 5990 | |||
| GenBank mRNA | AK125968.1 | - | 5 | 3086 | ||
| BX648636.1 | - | 7 | 3773 | |||
| CR749347.1 | - | 10 | 4613 | |||