siRNA Details
| Gene Symbol: LOC100133050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027503.1 | 4 | 688 | |||
| GenBank mRNA | BI830432.1 | - | 4 | 687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100133050| Gene Symbol: LOC100133050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027503.1 | 4 | 688 | |||
| GenBank mRNA | BI830432.1 | - | 4 | 687 | ||