siRNA Details
| Gene Symbol: PRSS30P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026864.2 | 2 | 2850 | |||
| GenBank mRNA | AK122946.1 | - | 2 | 2738 | ||
| BC041609.1 | - | 3 | 581 | |||
| BC042597.1 | - | 3 | 2765 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX117035.1 | - | 2 | 510 | Chr. 16 - 2839589 - 2840411 [-] on Build GRCh38 | |

PRSS30P
Genomic Map