siRNA Details
| Gene Symbol: PRSS30P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026864.2 | 1 | 2202 | |||
| GenBank mRNA | AK122946.1 | - | 1 | 2090 | ||
| BC042597.1 | - | 2 | 2117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS30P| Gene Symbol: PRSS30P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026864.2 | 1 | 2202 | |||
| GenBank mRNA | AK122946.1 | - | 1 | 2090 | ||
| BC042597.1 | - | 2 | 2117 | |||