siRNA Details
| Gene Symbol: COX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027941.2 | 5 | 1122 | |||
| NR_027942.2 | 5 | 1108 | ||||
| NR_135677.1 | 5 | 2711 | ||||
| XM_017024194.1 | XP_016879683.1 | 6 | 6909 | |||
| XM_017024195.1 | XP_016879684.1 | 6 | 6913 | |||
| XM_017024196.1 | XP_016879685.1 | 6 | 1202 | |||
| GenBank mRNA | AF044321.1 | - | 5 | 2578 | ||
| AK124809.1 | - | 1 | 2897 | |||

COX11
Genomic Map