siRNA Details
| Gene Symbol: COX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027941.2 | 4 | 903 | |||
| NR_027942.2 | 4 | 889 | ||||
| NR_135677.1 | 4 | 2492 | ||||
| XM_011524342.2 | XP_011522644.1 | 4 | 4731 | |||
| XM_017024192.1 | XP_016879681.1 | 4 | 4731 | |||
| XM_017024193.1 | XP_016879682.1 | 4 | 4731 | |||
| XM_017024194.1 | XP_016879683.1 | 4 | 884 | |||
| XM_017024195.1 | XP_016879684.1 | 4 | 884 | |||
| XM_017024196.1 | XP_016879685.1 | 4 | 884 | |||
| GenBank mRNA | AF044321.1 | - | 4 | 2359 | ||

COX11
Genomic Map