siRNA Details
| Gene Symbol: COX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027941.2 | 4, 5 | 1003 | |||
| NR_027942.2 | 4, 5 | 989 | ||||
| NR_135677.1 | 4, 5 | 2592 | ||||
| GenBank mRNA | AF044321.1 | - | 4, 5 | 2459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

COX11| Gene Symbol: COX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027941.2 | 4, 5 | 1003 | |||
| NR_027942.2 | 4, 5 | 989 | ||||
| NR_135677.1 | 4, 5 | 2592 | ||||
| GenBank mRNA | AF044321.1 | - | 4, 5 | 2459 | ||