siRNA Details
| Gene Symbol: LINC00935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029448.1 | 5 | 591 | |||
| NR_029449.1 | 4 | 506 | ||||
| GenBank mRNA | AK131005.1 | - | 4 | 506 | ||
| BC036043.1 | - | 4 | 481 | |||
| BC054859.1 | - | 5 | 573 | |||
| BC146870.1 | - | 4 | 488 | |||
| BC146877.1 | - | 4 | 488 | |||

LINC00935
Genomic Map