siRNA Details
| Gene Symbol: LINC00935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029448.1 | 5 | 551 | |||
| NR_029449.1 | 4 | 466 | ||||
| GenBank mRNA | AK131005.1 | - | 4 | 466 | ||
| BC036043.1 | - | 4 | 441 | |||
| BC054859.1 | - | 5 | 533 | |||
| BC146870.1 | - | 4 | 448 | |||
| BC146877.1 | - | 4 | 448 | |||

LINC00935
Genomic Map