siRNA Details
| Gene Symbol: LINC00935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029448.1 | 5 | 639 | |||
| NR_029449.1 | 4 | 554 | ||||
| GenBank mRNA | AK131005.1 | - | 4 | 554 | ||
| BC036043.1 | - | 4 | 529 | |||
| BC054859.1 | - | 5 | 621 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00935| Gene Symbol: LINC00935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029448.1 | 5 | 639 | |||
| NR_029449.1 | 4 | 554 | ||||
| GenBank mRNA | AK131005.1 | - | 4 | 554 | ||
| BC036043.1 | - | 4 | 529 | |||
| BC054859.1 | - | 5 | 621 | |||