siRNA Details
| Gene Symbol: DKFZP434K028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026882.1 | 2 | 287 | |||
| GenBank mRNA | AL117582.1 | - | 2 | 272 | ||
| BC021187.1 | - | 2 | 278 | |||
| DB452059.1 | - | 2 | 289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DKFZP434K028| Gene Symbol: DKFZP434K028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026882.1 | 2 | 287 | |||
| GenBank mRNA | AL117582.1 | - | 2 | 272 | ||
| BC021187.1 | - | 2 | 278 | |||
| DB452059.1 | - | 2 | 289 | |||