siRNA Details
| Gene Symbol: DKFZP434K028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026882.1 | 4 | 646 | |||
| GenBank mRNA | BC021187.1 | - | 4 | 637 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG718341.1 | - | 4 | 636 | Chr. 11 - 61754161 - 61757655 [-] on Build GRCh38 | |

DKFZP434K028
Genomic Map