siRNA Details
| Gene Symbol: PCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_015342.2 | 3 | 1690 | |||
| NR_132312.1 | 4 | 1855 | ||||
| NR_132313.1 | 3 | 1555 | ||||
| GenBank mRNA | AF103907.1 | - | LIT1552 DD3 |
4 | 1856 | |
| AY894120.1 | - | 3 | 1688 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF103908.3 | - | 1 | 3368 | Chr. 9 - 76782136 - 76787569 [+] on Build GRCh38 | |

PCA3
Genomic Map