siRNA Details
| Gene Symbol: FAM230C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027278.1 | 8 | 1112 | |||
| GenBank mRNA | AK097777.1 | - | 8 | 1052 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM230C| Gene Symbol: FAM230C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027278.1 | 8 | 1112 | |||
| GenBank mRNA | AK097777.1 | - | 8 | 1052 | ||