siRNA Details
| Gene Symbol: DKFZP434L187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026771.1 | 3 | 1834 | |||
| GenBank mRNA | AL117445.1 | - | 2 | 1724 | ||
| BC130408.1 | - | 3 | 1834 | |||
| BC130410.1 | - | 3 | 1834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DKFZP434L187| Gene Symbol: DKFZP434L187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026771.1 | 3 | 1834 | |||
| GenBank mRNA | AL117445.1 | - | 2 | 1724 | ||
| BC130408.1 | - | 3 | 1834 | |||
| BC130410.1 | - | 3 | 1834 | |||