siRNA Details
| Gene Symbol: DKFZP434A062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026964.3 | 1 | 4774 | |||
| GenBank mRNA | AK127730.1 | - | 1 | 4768 | ||
| AL080191.1 | - | 1 | 2677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DKFZP434A062| Gene Symbol: DKFZP434A062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026964.3 | 1 | 4774 | |||
| GenBank mRNA | AK127730.1 | - | 1 | 4768 | ||
| AL080191.1 | - | 1 | 2677 | |||