siRNA Details
| Gene Symbol: CYP2B7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001278.1 | 7 | 1062 | |||
| GenBank mRNA | DQ198366.1 | - | 7 | 1054 | ||
| DQ198367.1 | - | 7 | 1054 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP2B7P| Gene Symbol: CYP2B7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001278.1 | 7 | 1062 | |||
| GenBank mRNA | DQ198366.1 | - | 7 | 1054 | ||
| DQ198367.1 | - | 7 | 1054 | |||