siRNA Details
| Gene Symbol: CYP2B7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001278.1 | 2 | 312 | |||
| GenBank mRNA | AK307933.1 | - | 2 | 358 | ||
| AK310944.1 | - | 2 | 362 | |||
| DQ198366.1 | - | 2 | 304 | |||
| DQ198367.1 | - | 2 | 304 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP2B7P| Gene Symbol: CYP2B7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001278.1 | 2 | 312 | |||
| GenBank mRNA | AK307933.1 | - | 2 | 358 | ||
| AK310944.1 | - | 2 | 362 | |||
| DQ198366.1 | - | 2 | 304 | |||
| DQ198367.1 | - | 2 | 304 | |||