siRNA Details
| Gene Symbol: TOLLIP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029409.1 | 1 | 918 | |||
| GenBank mRNA | BC042808.1 | - | 1 | 799 | ||
| BC066355.1 | - | 1 | 723 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOLLIP-AS1| Gene Symbol: TOLLIP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029409.1 | 1 | 918 | |||
| GenBank mRNA | BC042808.1 | - | 1 | 799 | ||
| BC066355.1 | - | 1 | 723 | |||