siRNA Details
| Gene Symbol: TOLLIP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029409.1 | 1 | 355 | |||
| GenBank mRNA | BC042808.1 | - | 1 | 236 | ||
| BC066355.1 | - | 1 | 160 | |||
| DA359936.1 | - | 1 | 355 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI549672.1 | - | 1 | 241 | Chr. 11 - 1309827 - 1310691 [+] on Build GRCh38 | |

TOLLIP-AS1
Genomic Map