siRNA Details
| Gene Symbol: LINC00261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001558.3 | 2 | 168 | |||
| GenBank mRNA | BG655894.1 | - | 2 | 172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00261| Gene Symbol: LINC00261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001558.3 | 2 | 168 | |||
| GenBank mRNA | BG655894.1 | - | 2 | 172 | ||