siRNA Details
| Gene Symbol: LINC00261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001558.3 | 4 | 2368 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00261| Gene Symbol: LINC00261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001558.3 | 4 | 2368 | |||