siRNA Details
| Gene Symbol: MGC27382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027310.2 | 3, 4 | 713 | |||
| GenBank mRNA | BC015860.1 | - | 3, 4 | 694 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC27382| Gene Symbol: MGC27382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027310.2 | 3, 4 | 713 | |||
| GenBank mRNA | BC015860.1 | - | 3, 4 | 694 | ||