siRNA Details
| Gene Symbol: MGC27382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027310.2 | 3 | 506 | |||
| GenBank mRNA | AK091757.1 | - | 3 | 1143 | ||
| BC015860.1 | - | 3 | 487 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG536943.1 | - | 3 | 484 | Chr. 1 - 78229622 - 78342620 [+] on Build GRCh38 | |

MGC27382
Genomic Map