siRNA Details
| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026933.2 | 7 | 1325 | |||
| GenBank mRNA | DV080387.1 | - | 2 | 413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf173| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026933.2 | 7 | 1325 | |||
| GenBank mRNA | DV080387.1 | - | 2 | 413 | ||