siRNA Details
| Gene Symbol: C20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026933.2 | 8 | 1409 | |||
| GenBank mRNA | DV080387.1 | - | 3 | 497 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DW009790.1 | - | 5 | 706 | Chr. 20 - 35522517 - 35528255 [-] on Build GRCh38 | |

C20orf173
Genomic Map