siRNA Details
| Gene Symbol: OSTCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028496.1 | 3 | 885 | |||
| GenBank mRNA | BC024224.1 | - | 3 | 889 | 3998034 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OSTCP1| Gene Symbol: OSTCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028496.1 | 3 | 885 | |||
| GenBank mRNA | BC024224.1 | - | 3 | 889 | 3998034 | |