siRNA Details
| Gene Symbol: KIAA0125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026800.2 | 6 | 2891 | |||
| GenBank mRNA | AK125627.1 | - | 6 | 2892 | ||
| BC152473.1 | - | 1 | 1170 | |||
| D50915.1 | - | 1 | 1170 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA0125| Gene Symbol: KIAA0125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026800.2 | 6 | 2891 | |||
| GenBank mRNA | AK125627.1 | - | 6 | 2892 | ||
| BC152473.1 | - | 1 | 1170 | |||
| D50915.1 | - | 1 | 1170 | |||