siRNA Details
| Gene Symbol: LINC00899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027036.1 | 4 | 1214 | |||
| GenBank mRNA | AL137569.1 | - | 1 | 3755 | ||
| BC037213.1 | - | 3 | 2547 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX091020.1 | - | 1 | 656 | Chr. 22 - 46040267 - 46040958 [-] on Build GRCh38 | |

LINC00899
Genomic Map